Learn More
Escherichia coli O and K antigens, Rabbit, FITC, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Antigen | Escherichia coli O and K antigens |
| Applications | ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | In 10mM PBS, pH 7.2 (10mg/mL BSA, 0.09% sodium azide) |
| Host Species | Rabbit |
| Immunogen | Mixture of Escherichia coli serotypes. |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.