Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mycobacterium tuberculosis CFP10 (Rv3874), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Mycobacterium tuberculosis CFP10 (Rv3874).
Supplier: Abnova Corporation PAB1265
Specifications
Mycobacterium tuberculosis CFP10 (Rv3874) | |
Polyclonal | |
Rabbit polyclonal antibody raised against Mycobacterium tuberculosis CFP10 (Rv3874). | |
In PBS, pH 7.4 (0.09% sodium azide) | |
CFP10 (Rv3874) of Mycobacterium tuberculosis. | |
RUO | |
This antibody reacts with CFP10 (LHP), a 12.3 KDa antigen encoded by gene Rv3874 of Mycobacterium tuberculosis. This antibody recognizes CFP10 of Mycobacterium tuberculosis H37Rv and Mycobacterium bovis; it reacts also with recombinant antigen produced in Escherichia coli. | |
Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Rabbit | |
100 μg | |
Primary | |
Bacteria | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction