Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycobacterium tuberculosis EsaT-6 (Rv3875), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137526 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-526 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-526 Supplier Abnova Corporation Supplier No. PAB1266
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against Mycobacterium tuberculosis EsaT-6 (Rv3875).

Specifications

Antigen Mycobacterium tuberculosis EsaT-6 (Rv3875)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Mycobacterium tuberculosis EsaT-6 (Rv3875).
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Rabbit
Immunogen ESAT-6 (Rv3875) of Mycobacterium tuberculosis.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with EsaT-6, a 11.8 KDa antigen encoded by gene Rv3875 of Mycobacterium tuberculosis. This antibody recognizes EsaT-6 of Mycobacterium tuberculosis H37Rv and Mycobacterium bovis; it reacts also with recombinant antigen produced in Escherichia coli.
Target Species Bacteria
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.