Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pichia pastoris, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137805 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-805 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-805 Supplier Abnova Corporation Supplier No. PAB14023

Rabbit polyclonal antibody raised against Pichia pastoris.

Specifications

Antigen Pichia pastoris
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Rabbit
Immunogen Whole intact cells of Pichia pastoris.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity All antigens. Cross-reactive with Candida albicans (strong) and S. cerevisiae (weak).
Target Species Bacteria
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.