Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saccharomyces cerevisiae SUA7, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115257 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-257 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-257 Supplier Abnova Corporation Supplier No. PAB1165

Rabbit polyclonal antibody raised against full-length recombinant Saccharomyces cerevisiae SUA7.

Specifications

Antigen Saccharomyces cerevisiae SUA7
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant Saccharomyces cerevisiae SUA7.
Dilution The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Gene SUA7
Gene Alias SOH4
Gene Symbols SUA7
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length Saccharomyces cerevisiae SUA7.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 856201
Target Species Yeast
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.