Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saccharomyces cerevisiae TAF10, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115285 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-285 Supplier Abnova Corporation Supplier No. PAB1168

Rabbit polyclonal antibody raised against full-length recombinant Saccharomyces cerevisiae TAF10.

Specifications

Antigen Saccharomyces cerevisiae TAF10
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant Saccharomyces cerevisiae TAF10.
Dilution The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Gene TAF10
Gene Alias TAF23/TAF25
Gene Symbols TAF10
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length Saccharomyces cerevisiae TAF10.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 851745
Target Species Yeast
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.