Learn More
Saccharomyces cerevisiae TAF11, Rabbit, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Antigen | Saccharomyces cerevisiae TAF11 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyoclonal antibody raised against partial recombinant Saccharomyces cerevisiae TAF11. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | In buffer containing 0.09% sodium azide |
| Gene | TAF11 |
| Gene Alias | TAF40 |
| Gene Symbols | TAF11 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.