Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saccharomyces cerevisiae TAF4, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115266 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-266 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-266 Supplier Abnova Corporation Supplier No. PAB1166

Rabbit polyoclonal antibody raised against partial recombinant Saccharomyces cerevisiae TAF4.

Specifications

Antigen Saccharomyces cerevisiae TAF4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyoclonal antibody raised against partial recombinant Saccharomyces cerevisiae TAF4.
Dilution The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Gene TAF4
Gene Alias MPT1/TAF48/TSG2
Gene Symbols TAF4
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1-200 Saccharomyces cerevisiae TAF4.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 855017
Target Species Yeast
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.