Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Staphylococcus aureus Enterotoxin A, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137699 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-699 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-699 Supplier Abnova Corporation Supplier No. PAB13828

Rabbit polyclonal antibody raised against Staphylococcus aureus Enterotoxin A.

Specifications

Antigen Staphylococcus aureus Enterotoxin A
Applications Immunodiffusion
Classification Polyclonal
Conjugate Unconjugated
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Rabbit
Immunogen Purified Staphylococcus aureus Enterotoxin A.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Staphylococcus aureus Enterotoxin A. Non-reactive with other Staph. toxins.
Target Species Staphylococcus species
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.