Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IBSP, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116750 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-750 Supplier Abnova Corporation Supplier No. PAB13571
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native IBSP.

The protein encoded by this gene is a major structural protein of the bone matrix. It constitutes approximately 12% of the noncollagenous proteins in human bone and is synthesized by skeletal-associated cell types, including hypertrophic chondrocytes, osteoblasts, osteocytes, and osteoclasts. The only extraskeletal site of its synthesis is the trophoblast. This protein binds to calcium and hydroxyapatite via its acidic amino acid clusters, and mediates cell attachment through an RGD sequence that recognizes the vitronectin receptor. [provided by RefSeq

Specifications

Antigen IBSP
Applications ELISA, Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native IBSP.
Dilution Immunocytochemistry (1:100) The optimal working dilution should be determined by the end user.
Gene IBSP
Gene Alias BNSP/BSP/BSP-II/SP-II
Gene Symbols IBSP
Host Species Rabbit
Immunogen Native purified IBSP from human bone sialoprotein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3381
Test Specificity Recognizes human and rat bone sialoprotein. Does not cross-react with human osteopontin or osteonectin.
Target Species Human, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with distilled water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.