Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-ICAT, Polyclonal,

Catalog No. ABC939MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC939MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC939MI Supplier MilliporeSigma™ Supplier No. ABC939
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects ICAT in Human samples, and it is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting

Beta-catenin-interacting protein 1 (UniProt Q9NSA3; also known as Beta-catenin-interacting protein ICAT, Inhibitor of beta-catenin and Tcf-4) is encoded by the CTNNBIP1 (also known as ICAT) gene (Gene ID 56998) in human. Inhibitor of beta-catenin and TCF (ICAT) negatively regulates T-cell factor/lymphoid enhancer factor (TCF/LEF)–beta-catenin transcriptional activity by competing against TCF/LEF for beta-catenin binding. ICAT gene knockout leads to premature death of newborn mice with gut, kidney, and craniofacial anomalies, implicating an important role of ICAT in epithelial establishment/maintenance and neural crest cell differentiation. The motility of melanoma cells through 3D matrices in vitro relies on two interconvertible modes of movements. ICAT is reported to suppress the expression of developmentally downregulated 9 (NEDD9) of Wnt/beta-catenin signaling pathway in human Lu1205 melanoma cells, resulting in a shift from the Rac-dependent mesenchymal movement to the Rho/ROCK-dependent amoeboid motility in vitro and enhanced colonization of transplanted Lu1205 in the lungs of nude mice in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICAT
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. Q9NSA3
Gene Symbols CTNNBIP1; ICAT
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the C-terminus of human ICAT.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_064633
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.