Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICOS, Rabbit, Clone: SP98, Abnova™

Catalog No. 89108363 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-363 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-363 Supplier Abnova Corporation Supplier No. MAB10048

Rabbit monoclonal antibody raised against synthetic peptide of ICOS.

The protein encoded by this gene belongs to the CD28 and CTLA-4 cell-surface receptor family. It forms homodimers and plays an important role in cell-cell signaling, immune responses, and regulation of cell proliferation. [provided by RefSeq

Specifications

Antigen ICOS
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone SP98
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of ICOS.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene ICOS
Gene Alias AILIM/CD278/MGC39850
Gene Symbols ICOS
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human ICOS.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29851
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.