Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IDE, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89130836 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-836 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-836 Supplier Abnova Corporation Supplier No. PAB3109
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of IDE.

This gene may belong to a protease family responsible for intercellular peptide signalling. Though its role in the cellular processing of insulin has not yet been defined, insulin-degrading enzyme is thought to be involved in the termination of the insulin response. [provided by RefSeq

Specifications

Antigen IDE
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of IDE.
Dilution Western Blot (1:1000) Immunohistochemistry (1:50) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene IDE
Gene Alias FLJ35968/INSULYSIN
Gene Symbols IDE
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to internal region of human IDE.
Purification Method Protein A purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3416
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.