Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-IDH1, Polyclonal,

Catalog No. ABC1483MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC1483MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC1483MI Supplier MilliporeSigma™ Supplier No. ABC1483
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects IDH1 in Human samples, and it is validated for Western Blotting

Isocitrate dehydrogenase 1 (IDH1) is a 46kDa NADP-dependent enzyme, which catalyzes the decarboxylation of isocitrate into α-ketoglutarate. IDH1 may also play a role in the prevention of oxidative damage, and the shuttling of proteins to peroxisomes. It is widely reported that mutations in IDH1 result in multiple forms of gliomas. The Arginine to Serine substitution at amino acid 132 is seen in gliomas, abolishes magnesium binding and alters enzyme activity so that isocitrate is no longer converted to alpha-ketoglutarate but instead alpha-ketoglutarate is converted to R(-)-2-hydroxyglutarate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDH1
Applications Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing PBS (pH 7.2) with 0.02% sodium azide.
Gene Accession No. O75874
Gene Symbols IDH1; PICD
Host Species Rabbit
Immunogen IDH1 antibody was raised against a 13 amino acid synthetic peptide near the carboxy terminus of human IDH1.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.