Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-IFIT3, Polyclonal,

Catalog No. ABF1048MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABF1048MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF1048MI Supplier MilliporeSigma™ Supplier No. ABF1048
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects IFIT3 in Human, Mouse samples, and it is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting

Interferon-induced protein with tetratricopeptide repeats 3 (UniProt Q64345; also known as GARG-49, Glucocorticoid-attenuated response gene 49 protein. IFIT-3, IRG2, p49) is encoded by the Ifit3 (also known as Garg49, Ifi49, Isg49) gene (Gene ID 5959) in murine species. IFIT1, IFIT2, and IFIT3 constitute the family of interferon-stimulated genes (ISGs) that are induced in response to virus infection, stimulation by interferons (IFNs), or double-stranded RNA (dsRNA). The proteins encoded by IFIT1 and IFIT2 are called ISG56 (p56) and ISG54 (p54) in both human and murine species, while the protein encoded by IFIT3 is of a different size in murine (ISG49 or p49) than its human counterpart (ISG60 or p60). Studies conducted in murine species reveals that the three members exhibit distinct properties and induction patterns in different cell types. While all three murine proteins are shown to bind the c-subunit of the translation initiation factor eIF3, p49/Ifit3 does not inhibit protein synthesis as p56/Ifit1 and p54/Ifit2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFIT3
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. Q64345
Gene Symbols Ifit3; Garg49; Ifi49; Isg49
Host Species Rabbit
Immunogen His-tagged recombinant protein corresponding to mouse IFIT3.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_034631
Target Species Human, Mouse
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.