Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFNA2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120488 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-488 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-488 Supplier Abnova Corporation Supplier No. PAB18935

Rabbit polyclonal antibody raised against recombinant IFNA2.

Sequence: CDLPQTHSLGSRRTLMLLAQMRKISLFSCLKDRHDFGFPQEEFGNQFQKAETIPVLHEMIQQIFNLFSTKDSSAAWDETLLDKFYTELYQQLNDLEACVIQGVGVTETPLMNEDSILAVRKYFQRITLYLKEKKYSPCAWEVVRAEIMRSFSLSTNLQESLRSKE

Specifications

Antigen IFNA2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant IFNA2.
Dilution ELISA (1:500-1:1000) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene IFNA2
Gene Alias IFNA/INFA2/MGC125764/MGC125765
Gene Symbols IFNA2
Host Species Rabbit
Immunogen Recombinant protein corresponding to human IFNA2.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3440
Target Species Human
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with 100μL distilled water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.