Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFNAR1 (phospho S535/S539), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89134270 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-270 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-270 Supplier Abnova Corporation Supplier No. PAB9607
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of IFNAR1.

Specifications

Antigen IFNAR1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of IFNAR1.
Dilution Immunohistochemistry (1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA)
Gene IFNAR1
Gene Alias AVP/IFN-alpha-REC/IFNAR/IFNBR/IFRC
Gene Symbols IFNAR1
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S535/S539 of human IFNAR1.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3454
Target Species Bovine, Canine, Human, Mouse, Primate, Rat, Sheep
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.