Learn More
IGF1 Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of IGF1.
Supplier: Abnova Corporation PAB19107
Description
The protein encoded by this gene is similar to insulin in function and structure and is a member of a family of proteins involved in mediating growth and development. The encoded protein is processed from a precursor, bound by a specific receptor, and secreted. Defects in this gene are a cause of insulin-like growth factor I deficiency. Several transcript variants encoding different isoforms have been found for this gene
Specifications
IGF1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of IGF1. | |
Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
IGFI | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
3479 | |
Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1 ug/mL) The optimal working dilution should be determined by the end user. | |
IGF1 | |
IGF1 | |
A synthetic peptide corresponding to internal region of human IGF1. | |
100 μg | |
Primary | |
Human, Rat | |
Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only