Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF2R Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89120000 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-000 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-000 Supplier Abnova Corporation Supplier No. PAB18437
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of IGF2R.

The protein encoded by this gene is a receptor for both, insulin-like growth factor 2 (IGF2) and mannose 6-phosphate (M6P). The IGF2 and M6P binding sites are located on different segments of the receptor. This receptor functions in the intracellular trafficking of lysosomal enzymes, the activation of transforming growth factor beta, and the degradation of IGF2. In mice, the homologous gene is expressed only from the maternal chromosome. [provided by RefSeq

Specifications

Antigen IGF2R
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of IGF2R.
Dilution Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user.
Formulation In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
Gene IGF2R
Gene Alias CD222/CIMPR/M6P-R/MPR1/MPRI
Gene Symbols IGF2R
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human IGF2R.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3482
Test Specificity This antibody is specific to IGF2R.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.