Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGJ, Rabbit, Clone: SP105, Abnova™

Catalog No. 89108367 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-367 Supplier Abnova Corporation Supplier No. MAB10051

Rabbit monoclonal antibody raised against synthetic peptide of IGJ.

Specifications

Antigen IGJ
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone SP105
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of IGJ.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene IGJ
Gene Alias IGCJ/JCH
Gene Symbols IGJ
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region of human IGJ.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3512
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.