Learn More
IGSF11, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant IGSF11.
Supplier: Abnova Corporation PAB24502
Description
IGSF11 is an immunoglobulin (Ig) superfamily member that is preferentially expressed in brain and testis. It shares significant homology with coxsackievirus and adenovirus receptor (CXADR; MIM 602621) and endothelial cell-selective adhesion molecule (ESAM).[supplied by OMIM
Sequence: PPKCSSAKAFHTEISSSDNNTLTSSNAYNSRYWSNNPKVHRNTESVSHFSDLGQSFSFHSGNANIPSIYANGTHLVPGQHKTLVVTANRGSSPQVMSRSNGSVSRKPSpecifications
| IGSF11 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant IGSF11. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| BT-IgSF/CXADRL1/Igsf13/MGC35227/VSIG3 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 152404 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| IGSF11 | |
| IGSF11 | |
| Recombinant protein corresponding to amino acids of human IGSF11. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only