Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF8, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89127218 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-218 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-218 Supplier Abnova Corporation Supplier No. PAB25604
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of IGSF8.

Specifications

Antigen IGSF8
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of IGSF8.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene IGSF8
Gene Alias CD316/CD81P3/EWI2/PGRL
Gene Symbols IGSF8
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acids at internal region of human IGSF8.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93185
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins, except SIAH2 (53%).
Target Species Bovine, Human, Monkey, Pig, Rabbit
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.