Learn More
IKBKB Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
NFKB1 (MIM 164011) or NFKB2 (MIM 164012) is bound to REL (MIM 164910), RELA (MIM 164014), or RELB (MIM 604758) to form the NFKB complex. The NFKB complex is inhibited by I-kappa-B proteins (NFKBIA, MIM 164008, or NFKBIB, MIM 604495), which inactivate NF-kappa-B by trapping it in the cytoplasm. Phosphorylation of serine residues on the I-kappa-B proteins by kinases (IKBKA, MIM 600664, or IKBKB) marks them for destruction via the ubiquitination pathway, thereby allowing activation of the NF-kappa-B complex. Activated NFKB complex translocates into the nucleus and binds DNA at kappa-B-binding motifs such as 5-prime GGGRNNYYCC 3-prime or 5-prime HGGARNYYCC 3-prime (where H is A, C, or T; R is an A or G purine; and Y is a C or T pyrimidine).[supplied by OMIM
Specifications
Specifications
| Antigen | IKBKB |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic peptide of IKBKB. |
| Dilution | Immunofluorescence (1:100-1:200) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) |
| Gene | IKBKB |
| Gene Alias | FLJ40509/IKK-beta/IKK2/IKKB/MGC131801/NFKBIKB |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.