Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKBKE Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89116134 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-134 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-134 Supplier Abnova Corporation Supplier No. PAB12855
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of IKBKE.

Sequence: NRIIERLNRVPAPPDV

Specifications

Antigen IKBKE
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of IKBKE.
Dilution Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene IKBKE
Gene Alias IKK-i/IKKE/IKKI/KIAA0151/MGC125294/MGC125295/MGC125297
Gene Symbols IKBKE
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 701-716 of human IKBKE.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9641
Test Specificity A 80 KDa band should be detected. It has no cross response to IKKa, IKKb, or IKKy.
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.