Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKK-alpha/beta (phospho S176), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of IKK-alpha/beta.
Supplier: Abnova Corporation PAB16976
Specifications
IKK-alpha/beta | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of IKK-alpha/beta. | |
In 20mM PBS, 0.15M NaCl, pH 7.2 (0.01% sodium azide) | |
Synthetic phosphopeptide corresponding to residues surrounding S176 of human IKK-alpha/beta. | |
RUO | |
This antibody detects endogenous levels of IKK-alpha/IKK-beta only when phosphorylated at serine 180/181. | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:4000) The optimal working dilution should be determined by the end user. | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction