Learn More
Description
Specifically detects IKK beta in Human samples, and it is validated for Immunocytochemistry, Western Blotting
Specifications
Specifications
| Antigen | IKK beta |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Formulation | Purified rabbit polyclonal in buffer containing PBS (pH 7.2) with 0.02% sodium azide. |
| Gene Accession No. | O14920 |
| Gene Symbols | IKBKB; IKKB |
| Host Species | Rabbit |
| Immunogen | IKK beta antibody was raised against a peptide corresponding to amino acids near the carboxy terminus of human IKK beta (Genbank accession NoO14920), which differs from corresponding murine sequence by one amino acid. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.