Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-IL-21, Polyclonal,

Catalog No. ABF184MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABF184MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF184MI Supplier MilliporeSigma™ Supplier No. ABF184
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects IL-21 in Human samples, and it is validated for Flow Cytometry, Western Blotting

Interleukin-21 (UniProt Q9HBE4; also known as IL-21, Za11) is encoded by the IL21 (also known as CVID11) gene (Gene ID 59067) in human. IL-21 is a type I four-alpha-helical bundle cytokine primarily produced by CD4+ T cells and NKT cells. The heterodimeric receptor IL-21R is dominantly expressed by B cells and shares the common cytokine receptor gamma chain (γc) with receptors for IL-2, IL-4, IL-7, IL-9, and IL-15. Studies indicate that IL-21 potently regulates both adaptive and innate immune responses and possesses widely pleiotropic functions. IL-21 has also been linked to autoimmune disorders, allergies, inflammatory diseases, as well as the pathogenesis of cancer. Human IL-21 is initially translated with an N-terminal 22 aa signal peptide sequence, which is later removed to produce the 133 aa mature IL-21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-21
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9HBE4
Gene Symbols IL21
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human IL-21 sequence near the C-terminus.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_068575
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.