Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-IL-33, Polyclonal,

Catalog No. ABF108MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABF108MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF108MI Supplier MilliporeSigma™ Supplier No. ABF108
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects IL-33 in Mouse samples, and it is validated for Flow Cytometry, Western Blotting

IL-33 is a proinflammatory protein that shares structural and functional characteristics with the IL-1 cytokine family. It binds and signals through the IL-1RL1/ST2 receptor, leading to the activation of MAP kinases and NF-kappaB. IL-33 induces production of helper T-cell type 2 (TH2) cytokines, including IL-4, IL-5 and IL-13, and is a known mediator of many inflammation responses. Anti-IL-33, is an affinity purified rabbit polyclonal antibody that recognizes epitopes in the C-terminal half of murine IL-33. Reactivity to rat or other species has not been tested.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-33
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q8BVZ5
Gene Symbols Il33
Host Species Rabbit
Immunogen His-tagged recombinant protein corresponding to the C-Terminal of mouse IL-33.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_598536
Target Species Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.