Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL17RD, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89118069 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-069 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-069 Supplier Abnova Corporation Supplier No. PAB16178
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant IL17RD.

Fibroblast growth factors (FGFs; see MIM 603726) are secreted proteins involved in cellular proliferation, migration, differentiation, and survival. FGF activity is negatively regulated by members of the 'sprouty' family (e.g., SPRY1, MIM 602465). The SEF protein is a modulator of FGF signaling.[supplied by OMIM

Specifications

Antigen IL17RD
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant IL17RD.
Dilution ELISA (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS.
Gene IL17RD
Gene Alias DKFZp434N1928/FLJ35755/IL-17RD/IL17RLM/MGC133309/SEF
Gene Symbols IL17RD
Host Species Rabbit
Immunogen Recombinant protein corresponding to human IL17RD.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54756
Target Species Human
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.