Learn More
IL17RE Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant IL17RE.
Supplier: Abnova Corporation PAB21053
Description
This gene encodes a protein that shares limited similarity with the interleukin-17 receptor. The function of the encoded protein has not yet been determined. Three alternatively spliced transcript variants of this gene encoding two distinct isoforms have been reported. [provided by RefSeq
Sequence: EKSHHISIPSPDISHKGLRSKRTQPSDPETWESLPRLDSQRHGGPEFSFDLLPEARAIRVTISSGPEVSVRLCHQWALECEELSSPYDVQKIVSGGHTVELPYEFLLPCLCMEASYLQEDTVRRKKCPFQSWPEAYGSDFWKSVHFTDYSSpecifications
IL17RE | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant IL17RE. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
FLJ23658/MGC71884 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
132014 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
IL17RE | |
IL17RE | |
Recombinant protein corresponding to amino acids of human IL17RE. | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only