Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Il18 Rabbit anti-Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Il18.
Supplier: Abnova Corporation PAB19936
Specifications
Il18 | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant Il18. | |
In 20mM KH2PO4, 150mM NaCl, pH 7.2 | |
Igif/Il-18 | |
Rabbit | |
Protein A purification | |
RUO | |
16173 | |
Mouse | |
Liquid |
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
ELISA (1:1000-1:5000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
Il18 | |
Il18 | |
Recombinant protein corresponding to a 155 amino acids fragment of mouse Il18. | |
500 μg | |
Primary | |
This antibody is primarily directed against mature 18,000 MW mouse IL-18 and is useful in determining its presence in various assays. This antibody will also recognize the 24,000 inactive precursor form of mouse IL-18. In general, this antibody also detects rat IL-18 in the same formats using similar dilutions. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction