Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL1B Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89119864 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-864 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-864 Supplier Abnova Corporation Supplier No. PAB18296
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of IL1B.

The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine is produced by activated macrophages as a proprotein, which is proteolytically processed to its active form by caspase 1 (CASP1/ICE). This cytokine is an important mediator of the inflammatory response, and is involved in a variety of cellular activities, including cell proliferation, differentiation, and apoptosis. The induction of cyclooxygenase-2 (PTGS2/COX2) by this cytokine in the central nervous system (CNS) is found to contribute to inflammatory pain hypersensitivity. This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. [provided by RefSeq

Specifications

Antigen IL1B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of IL1B.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal)
Gene IL1B
Gene Alias IL-1/IL1-BETA/IL1F2
Gene Symbols IL1B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids at C-terminus of human IL1B.
Purification Method Immunoaffinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3553
Target Species Human
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.