Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL20RA Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89114162 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-162 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-162 Supplier Abnova Corporation Supplier No. PAB0346
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of IL20RA.

The protein encoded by this gene is a receptor for interleukin 20 (IL20), a cytokine that may be involved in epidermal function. The receptor of IL20 is a heterodimeric receptor complex consisting of this protein and interleukin 20 receptor beta (IL20B). This gene and IL20B are highly expressed in skin. The expression of both genes is found to be upregulated in Psoriasis. [provided by RefSeq

Specifications

Antigen IL20RA
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of IL20RA.
Dilution The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.05% sodium azide, 0.2% gelatin
Gene IL20RA
Gene Alias FLJ40993/IL-20R1/ZCYTOR7
Gene Symbols IL20RA
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 159-175 of human IL20RA .
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53832
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.