Learn More
IL21R Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of IL21R.
Supplier: Abnova Corporation PAB17987
Description
The protein encoded by this gene is a cytokine receptor for interleukin 21 (IL21). It belongs to the type I cytokine receptors, and has been shown to form a heterodimeric receptor complex with the common gamma-chain, a receptor subunit also shared by the receptors for interleukin 2, 4, 7, 9, and 15. This receptor transduces the growth promoting signal of IL21, and is important for the proliferation and differentiation of T cells, B cells, and natural killer (NK) cells. The ligand binding of this receptor leads to the activation of multiple downstream signaling molecules, including JAK1, JAK3, STAT1, and STAT3. Knockout studies of a similar gene in mouse suggest a role for this gene in regulating immunoglobulin production. Three alternatively spliced transcript variants encoding the same protein have been described. [provided by RefSeq
Specifications
IL21R | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of IL21R. | |
In PBS pH 7.2 (50% glycerol, 1% BSA, 0.02% sodium azide) | |
MGC10967/NILR | |
Rabbit | |
100 μg | |
Primary | |
human A431 cell lysates can be used as a positive control and an approximately 60 KDa band should be detected. | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
IL21R | |
IL21R | |
A synthetic peptide corresponding to amino acids 20-32 of human IL21R. | |
RUO | |
50615 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only