Learn More
IL27 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of IL27.
Supplier: Abnova Corporation PAB13199
Description
The protein encoded by this gene is one of the subunits of a heterodimeric cytokine complex. This protein is related to interleukin 12A (IL12A). It interacts with Epstein-Barr virus induced gene 3 (EBI3), a protein similar to interleukin 12B (IL12B), and forms a complex that has been shown to drive rapid expansion of naive but not memory CD4(+) T cells. The complex is also found to synergize strongly with interleukin 12 to trigger interferon gamma (IFNG) production of naive CD4(+) T cells. The biological effects of this cytokine are mediated by the class I cytokine receptor (WSX1/TCRR). [provided by RefSeq]
Specifications
IL27 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of IL27. | |
In PBS (0.02% sodium azide) | |
IL-27/IL-27A/IL27p28/IL30/MGC71873/p28 | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
Immunocytochemistry, Western Blot | |
Unconjugated | |
Western Blot (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
IL27 | |
IL27 | |
A synthetic peptide corresponding to N-terminus 16 amino acids of human IL27. | |
RUO | |
246778 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only