Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL7, Rabbit, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89121443 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-443 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-443 Supplier Abnova Corporation Supplier No. PAB19935
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant IL7.

The protein encoded by this gene is a cytokine important for B and T cell development. This cytokine and the hepatocyte growth factor (HGF) form a heterodimer that functions as a pre-pro-B cell growth-stimulating factor. This cytokine is found to be a cofactor for V(D)J rearrangement of the T cell receptor beta (TCRB) during early T cell development. This cytokine can be produced locally by intestinal epithelial and epithelial goblet cells, and may serve as a regulatory factor for intestinal mucosal lymphocytes. Knockout studies in mice suggested that this cytokine plays an essential role in lymphoid cell survival. [provided by RefSeq

Specifications

Antigen IL7
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
Description Rabbit polyclonal antibody raised against full length recombinant IL7.
Dilution ELISA (1:20000-1:100000) Western Blot (1:2000-1:10000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene IL7
Gene Alias IL-7
Gene Symbols IL7
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length human IL7.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3574
Test Specificity In ELISA and other immunoreactive assays, this antibody will recognize both native and recombinant human IL-7 in cell supernatants and certain body fluids. A control of similarly diluted normal rabbit IgG is recommended.
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.