Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Il8rb, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Il8rb.
Supplier: Abnova Corporation PAB8942
Specifications
Il8rb | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Il8rb. | |
In buffer containing 0.02% sodium azide | |
Il8rb | |
A synthetic peptide corresponding to C-terminus of rat/mouse Il8rb. | |
RUO | |
12765/29385 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (1:500) Immunoprecipitation (1:250) The optimal working dilution should be determined by the end user. | |
CD128/CDw128/CXCR2/Cmkar2/Gpcr16/IL-8Rh/IL-8rb/IL8RA/mIL-8RH | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction