Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILF3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89130044 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-044 Supplier Abnova Corporation Supplier No. PAB28534
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant ILF3.

This gene encodes a double-stranded RNA (dsRNA) binding protein that complexes with other proteins, dsRNAs, small noncoding RNAs, and mRNAs to regulate gene expression and stabilize mRNAs. This protein was first discovered to be a subunit of the nuclear factor of activated T-cells (NFAT); a transcription factor required for T-cell expression of interleukin 2. NFAT is a heterodimer of 45kDa and 90kDa proteins, the larger of which is the product of this gene. These proteins have been shown to affect the redistribution of nuclear mRNA to the cytoplasm. Knockdown of NF45 or NF90 protein retards cell growth; possibly by inhibition of mRNA stabilization. In contrast, an isoform (NF110) of this gene that is predominantly restricted to the nucleus has only minor effects on cell growth when its levels are reduced. Alternative splicing results in multiple transcript variants encoding distinct isoforms

Sequence: VMAKHSSVYPTQEELEAVQNMVSHTERALKAVSDWIDEQEKGSSEQAESDNMDVPPEDDSKEGAGEQKTEHMTRTLRGVMRVGLVAKGLLLKGDLDLELVLLCKEKPTTALLDKVADNLAIQLAAVTEDKYEILQSVDDAAIVIK

Specifications

Antigen ILF3
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant ILF3.
Dilution Western Blot (1:100-1:500) Immunofluorescence (1-4 ug/ml) Immunohistochemistry (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
Gene ILF3
Gene Alias CBTF/DRBF/DRBP76/MMP4/MPHOSPH4/MPP4/NF-AT-90/NF110/NF90/NFAR/NFAR-1/NFAR2/TCP110/TCP80
Gene Symbols ILF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human ILF3.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3609
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.