Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Immunodeficiency Virus (HIV) P24, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137956 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-956 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-956 Supplier Abnova Corporation Supplier No. PAB14679
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant human Immunodeficiency Virus (HIV) P24.

Specifications

Antigen Immunodeficiency Virus (HIV) P24
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant human Immunodeficiency Virus (HIV) P24.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 20mM PBS, pH 7.4
Host Species Rabbit
Immunogen Recombinant protein corresponding to human Immunodeficiency Virus (HIV) P24.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes both native and recombinant HIV-P24.
Target Species Human, Virus
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.