Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INA Rabbit anti-Human, Mammals, Polyclonal Antibody, Abnova™

Catalog No. 89115603 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-603 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-603 Supplier Abnova Corporation Supplier No. PAB12106

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit polyclonal antibody raised against recombinant Ina.

Specifications

Antigen INA
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant Ina.
Dilution Immunofluorescence (1:500-1:1000) Western Blot (1:5000-1:10000) The optimal working dilution should be determined by the end user.
Formulation In 10mM sodium phosphate buffer, 250mM NaCl, pH 7.6 (15mg/mL BSA, 0.05% sodium azide, 0.01% thimerosal)
Gene Ina
Gene Alias Inexa/Intlaa/Nf66
Gene Symbols Ina
Host Species Rabbit
Immunogen Recombinant protein corresponding to rat Ina.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24503
Test Specificity Specifically recognizes a-internexin.
Target Species Human, Mammalia
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.