Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INAD, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89126378 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-378 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-378 Supplier Abnova Corporation Supplier No. PAB24713
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of inaD.

Specifications

Antigen INAD
Applications Dot Blot, ELISA, Western Blot
Classification Polyclonal
Concentration 0.55-0.62 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of inaD.
Dilution ELISA (1:20000-1:50000) Dot blot (1:20000-1:50000) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine buffer, pH 7.4 (stabilizing proteins, cryogenic stabilizers, glycerol, 0.02% sodium azide)
Gene inaD
Gene Alias CG3504/Ina-D
Gene Symbols inaD
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 659-674 of fruit fly inaD.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 37629
Target Species Fruitfly
Content And Storage Store at -20°C
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.