Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabbit anti-iNOS, Clone: K13-A, Novus Biologicals™

Rabbit Monoclonal Antibody has been used in 7 publications
Supplier: Novus Biologicals NBP1337800.1ML
Description
iNOS Monoclonal specifically detects iNOS in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
iNOS | |
Monoclonal | |
Unconjugated | |
20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide | |
EC 1.14.13.39, Hepatocyte NOS, HEP-NOSPeptidyl-cysteine S-nitrosylase NOS2, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS, type II, NOS2ANOS type II | |
Rabbit | |
Affinity Purified | |
RUO | |
4843 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
K13-A | |
ELISA 1:20000-1:50000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100-1:200 | |
P35228 | |
NOS2 | |
Peptide derived from the human iNOS sequence. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction