Learn More
Rabbit anti-Insect Cell Line SF21, Polyclonal, Abnova™
Supplier: Abnova Corporation PAB13994
Specifications
| Insect Cell Line SF21 | |
| Polyclonal | |
| In PBS | |
| Native purified from the ovarian derived cell line of Spodoptera frugiperda. | |
| Primary | |
| Purified |
| Dot Blot, ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| 1 mL | |
| Insect |