Learn More
IPO8, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant IPO8.
Supplier: Abnova Corporation PAB0302
Description
The importin-alpha/beta complex and the GTPase Ran mediate nuclear import of proteins with a classical nuclear localization signal. The protein encoded by this gene is a member of a class of approximately 20 potential Ran targets that share a sequence motif related to the Ran-binding site of importin-beta. This protein binds to the nuclear pore complex and, along with RanGTP and RANBP1, inhibits the GAP stimulation of the Ran GTPase. [provided by RefSeq
Specifications
IPO8 | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant IPO8. | |
In PBS (0.2% gelatin, 0.05% sodium azide) | |
FLJ26580/RANBP8 | |
Rabbit | |
100 μg | |
Primary | |
Chimpanzee, Human, Monkey | |
Liquid |
Immunocytochemistry, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
IPO8 | |
IPO8 | |
Recombinant protein corresponding to amino acids 850-900 of human IPO8. | |
RUO | |
10526 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only