Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IQSEC1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant IQSEC1.
Supplier: Abnova Corporation PAB23229
Description
Sequence: PSSETGTSLDSPSAYPQGPLVPGSSLSPDHYEHTSVGAYGLYSGPPGQQQRTRRPKLQHSTSILRKQAESpecifications
IQSEC1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant IQSEC1. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
ARFGEP100/KIAA0763 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
9922 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunofluorescence, Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
IQSEC1 | |
IQSEC1 | |
Recombinant protein corresponding to amino acids of human IQSEC1. | |
100 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction