Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRAK2 Rabbit anti-Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Irak2.
Supplier: Abnova Corporation PAB13162
Specifications
IRAK2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Irak2. | |
In PBS (0.02% sodium azide) | |
6330415L08Rik/AI649099/IRAK-2/MGC102586 | |
Rabbit | |
100 μg | |
Primary | |
This antibody is common to all four isoforms. Anti-mIRAK2 is mouse specific. Anti-IRAK2 has no cross response to IRAK. | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
Irak2 | |
Irak2 | |
A synthetic peptide corresponding to C-terminus 16 amino acids of mouse Irak2. | |
RUO | |
108960 | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction