Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IREB2, Rabbit, DyLight 488, Polyclonal Antibody, Abnova™

Catalog No. 89117404 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-404 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-404 Supplier Abnova Corporation Supplier No. PAB15013
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Ireb2.

Specifications

Antigen IREB2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Description Rabbit polyclonal antibody raised against synthetic peptide of Ireb2.
Dilution Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium borate
Gene Ireb2
Gene Alias D9Ertd85e/DKFZp564D1164/Irp2
Gene Symbols Ireb2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 100-200 of mouse Ireb2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64602
Test Specificity This antibody reacts with mouse IRP2 protein. Inital results have shown poor reactivity against human protein. The immunogen used for this antibody production has 94% homology with human and rat proteins. Rabbit Polyclonal to IRP2 By Western blot a band at ∼105 KDa is seen. Note that in brain lysates it has been reported that IRP2 appears to be degraded by low concentrations of free iron. Addition of 50 uM deferoxamine to the lysis buffer to chelate free iron appears to prevent this degradation.
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.