Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IREB2 Rabbit anti-Human, Mouse, Primates, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115635 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-635 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-635 Supplier Abnova Corporation Supplier No. PAB12137
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Ireb2.

Specifications

Antigen IREB2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Ireb2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Gene Ireb2
Gene Alias D9Ertd85e/DKFZp564D1164/Irp2
Gene Symbols Ireb2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 100-200 of mouse Ireb2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64602
Target Species Human, Mouse, Primate, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.