Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF4, Rabbit, Clone: SP114, Abnova™

Catalog No. 89108373 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-373 Supplier Abnova Corporation Supplier No. MAB10057

Rabbit monoclonal antibody raised against synthetic peptide of IRF4.

Specifications

Antigen IRF4
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone SP114
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of IRF4.
Dilution Western Blot (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene IRF4
Gene Alias LSIRF/MUM1
Gene Symbols IRF4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human IRF4.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3662
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.