Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF8 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89130056 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-056 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-056 Supplier Abnova Corporation Supplier No. PAB28546
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant IRF8.

Interferon consensus sequence-binding protein (ICSBP) is a transcription factor of the interferon (IFN) regulatory factor (IRF) family. Proteins of this family are composed of a conserved DNA-binding domain in the N-terminal region and a divergent C-terminal region that serves as the regulatory domain. The IRF family proteins bind to the IFN-stimulated response element (ISRE) and regulate expression of genes stimulated by type I IFNs, namely IFN-alpha and IFN-beta. IRF family proteins also control expression of IFN-alpha and IFN-beta-regulated genes that are induced by viral infection. [provided by RefSeq

Sequence: QGVFVKRLCQGRVFCSGNAVVCKGRPNKLERDEVVQVFDTSQFFRELQQFYNSQGRLPDGRVVLCFGEEFPDMAPLRSKLILVQIEQLYVRQLAEEAGKSCGAGSVMQAPEEPPPDQVFRMFPDICASHQRSFFRENQQIT

Specifications

Antigen IRF8
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant IRF8.
Dilution Immunohistochemistry(-) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
Gene IRF8
Gene Alias H-ICSBP/ICSBP/ICSBP1/IRF-8
Gene Symbols IRF8
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of recombinant IRF8.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3394
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.